Learn More
Graham Professional™ Tourniquet
- Textured surface eliminates slipping and pinching
- Tear resistant and powder free, with superior stretch and recovery
Specifications
Color
White
Latex Free
Yes
Product Type
Tourniquet
For Use With (Application)
Textured surface eliminates slipping and pinching
Disposable
Single-use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.