Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grams Iodine Solution, Certified, 0.025N ±0.001N, LabChem™

Catalog No. p-5389154
Click to view available options
Quantity:
500 mL
1 L
4 L

Chemical Identifiers

CAS 7732-18-5, 7681-11-0, 7553-56-2, 7647-01-0
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide

Specifications

Physical Form Liquid
CAS Max % 99.02, 0.64, 0.32, 0.02
Packaging Amber Glass
Solubility Soluble in water
Decomposition Information Iodine vapor; Potassium oxide
Color Red to Brown
Quantity 500 mL
Linear Formula I2
Formula Weight 253.809
Grade Certified
Concentration or Composition (by Analyte or Components) 0.025N ±0.001N
Chemical Name or Material Grams Iodine Solution
Show More Show Less
Hazard Statement GHS H Statement
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.