Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Graphene Oxide, TCI America™
SDP

Catalog No. G0443100MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg

Specifications

Color Black-Yellow
Quantity 100 mg
Physical Form Crystalline Powder
Chemical Name or Material Graphene Oxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.