Learn More
Graphite rod, 6.15mm (0.242in) dia x 152mm (6in) long, 99.9995% (metals basis), Thermo Scientific Chemicals
Description
It is used in various industries. Used in making. Anodes, Electrodes, Heating elements, Crucibles, Hot press dies, Welding plugs, Stirring rods, Lubrication sticks, Bushings & bearings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Chemical Name or Material | Graphite rod |
Packaging | Tube |
Quantity | 50 Pc. |
MDL Number | MFCD00144065 |
Solubility Information | Insoluble in water. |
Percent Purity | 99.9995% |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.