Learn More
Glass Expansion Graphite Sampler Cone Gasket
Supplier:  Glass Expansion TG5001
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Graphite Sampler Cone Gasket (3004382). Package of 1 graphite gasket for Thermo/VG Q, RQ, TX and X-Series ICP-MS sampler cones. Use a new gasket each time the cone is installed, even if it is the same cone, to ensure a proper seal. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When cones are not in use, keep them stored in original packaging for protection.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2930
missing translation for 'QuantityLeft'