Learn More
CODAN USA GREEN STRIP MINIBOR SET 50CS
Supplier:  CODAN USA BC261 TUBING
Encompass_Preferred
Green Strip MiniBore Extension Set, pressure-activated valve, slide clamp, male luer-lock 50/CS
Catalog No. NC1799946
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.