Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRI 977143, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective lysophosphatidic acid 2 (LPA2) receptor non-lipid agonist (EC50 = 3.3 μM). Does not activate other LPA GPCRs at concentrations up to 10 μM. Antiapoptotic; inhibits activation of caspases 3, 7, 8 and 9, Bax translocation and PARP-1 cleavage. Activates ERK1/2 survival pathway.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 391.44 |
Percent Purity | >98% |
Chemical Name or Material | GRI 977143 |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction