Learn More
Electron Microscopy Sciences Gilder Finder Grid F1, 300 mesh Gold Grid
Supplier:  Electron Microscopy Sciences G300F1AU
Encompass_Preferred
Used to identify specific areas of mesh by an alphabetical or numerical code built into the mesh. 3.05 mm diameter, 0.7 mil (18 um) thickness. The block of 625 (25x25) individual cells is divided into smaller blocks bounded by thicker crenellated bars. Each block of 25 cells has a central alphabet letter from A to Y for identification purposes.
Grid: Gold
Catalog No. 50-192-7776
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.