Learn More
Electron Microscopy Sciences Grid Holder for CPD
Supplier: Electron Microscopy Sciences 7019201
Machined from solid steel; has a stainless steel mesh on the top and the bottom. Featuring the most advanced design which assures no damage to the grids during processing. These holders are Brass-Nickel plated, very sturdy, and withstand all critical conditions. Holds 12 grids in numbered slots. Dimensions: 24 mm(D)x24 mm(H)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.