Learn More
Electron Microscopy Sciences Gilder Aperture Grid with Single 200 µm hole, Copper
Supplier:  Electron Microscopy Sciences GA200CU
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 200 um.
Material: Copper
Catalog No. 50-190-1287
missing translation for 'QuantityLeft'