Learn More
WELOC USA LLC WeLoc Gripper PA 150 Red
This item has a minimum order quantity of 3 per supplier requirementsThe WeLoc Gripper PA is the most powerful WeLoc closure. It is manufactured from DuPont’s Nylon, reinforced with glass fibre which further enhances its strength and durability.