Learn More
AdipoGen GS-441524 Remdesivir Metabol
Supplier: AdipoGen AGCR13722M050
Chemical. CAS 1191237-69-0. Formula C12H13N5O4. MW 291.3. GS-441524 is the active prodrug of the broad-spectrum antiviral Remdesivir. GS-441524 is an adenosine nucleotide analog in viral RNA production. It is a nucleotide analog inhibitor of RNA-dependent RNA polymerases RdRps. The compound interferes with the action of viral RNA polymerase and evades proofreading by viral exoribonuclease ExoN, causing a decrease in viral RNA production RNA synthesis arrest, either by terminating RNA chains or causing mutations. It inhibits MERS-CoV or SARS-CoV-infected HAE cultures EC50=74nM and 69nM and murine hepatitis virus MHV EC50=30nM. In vivo, remdesivir 25 and 50 mg/kg reduces lung viral titers and prevents weight loss in a mouse model of SARS-CoV infection.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.