Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSK 0660, Tocris Bioscience™

Catalog No. p-4817132 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective PPARδ antagonist

Selective PPARδ antagonist (IC50 values are 0.155, > 10 and ≥ 10 μM at PPARδ, PPARα and PPARγ respectively). Exhibits inverse agonist effects when administered by itself.

Chemical Identifiers

CAS 1014691-61-2
Molecular Formula C19H18N2O5S2
Molecular Weight (g/mol) 418.48
MDL Number MFCD12828770
InChI Key NDFKBGWLUHKMFY-UHFFFAOYSA-N
Synonym methyl 3-4-anilino-2-methoxyphenyl sulfamoyl thiophene-2-carboxylate, 3-2-methoxy-4-phenylamino phenyl amino sulfonyl-2-thiophenecarboxylicacidmethylester, 3-2-methoxy-4-phenylamino phenyl amino sulfonyl-2-thiophenecarboxylic acid methyl ester, methyl 3-n-2-methoxy-4-phenylamino phenyl sulfamoyl thiophene-2-carboxylate, 2-thiophenecarboxylic acid, 3-2-methoxy-4-phenylamino phenyl amino sulfonyl-, methyl ester, d06vbq, gsk hplc, methyl 3-4-anilino-2-methoxy-phenyl sulfamoyl thiophene-2-carboxylate
PubChem CID 46233311
IUPAC Name methyl 3-{[2-methoxy-4-(phenylamino)phenyl]sulfamoyl}thiophene-2-carboxylate
SMILES COC(=O)C1=C(C=CS1)S(=O)(=O)NC1=C(OC)C=C(NC2=CC=CC=C2)C=C1

Specifications

Quantity 10 mg
Formula Weight 418.49
Percent Purity >99%
Chemical Name or Material GSK 0660
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.