Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ GSK 1702934A
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
GSK 1702934A is a potent and selective TRPC3/6 activator (EC50 = 80 and 440 nM for TRPC3 and 6, respectively). Exhibits no activity at TRPV4, TRPA1, M1, M4, CaV1.2, hERG, NaV1.5, or CXCR5 receptors at concentrations <10 μmol/L. Induces TRPC3/6-currents in HEK293 cells transduced with recombinant human TRPC3/6. Directly activates TRPC channels through by-passing phospholipase-C signaling. Also induces calcium signals in HEK-TRPC6 cells (pEC50 = 6.6).
Specifications
Specifications
Chemical Name or Material | 1,3-Dihydro-1-[1-[(5,6,7,8-tetrahydro-4H-cyclohepta[b]thien-2-yl)carbonyl]-4-piperidinyl]-2H-benzimidazol-2-one |
CAS | 924377-85-5 |
Quantity | 10 mg |
Molecular Formula | C22H25N3O2S |
Formula Weight | 395.52 |
Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction