Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSK 4716, Tocris Bioscience™

Catalog No. p-4783999 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective ERRγ and ERRβ agonist

Selective agonist at estrogen-related receptors ERRβ and ERRγ. Displays selectivity over ERRα and the classical estrogen receptors.

Chemical Identifiers

CAS 101574-65-6
Molecular Formula C17H18N2O2
Molecular Weight (g/mol) 282.34
MDL Number MFCD00567155
InChI Key IKPPIUNQWSRCOZ-WOJGMQOQSA-N
Synonym e-4-hydroxy-n'-4-isopropylbenzylidene benzohydrazide, 4-hydroxy-benzoic acid 4-isopropyl-benzylidene-hydrazide, 4-hydroxy-2-1e-4-1-methylethyl phenyl methylene hydrazide, 2-4-1-methylethyl phenyl methylene hydrazide 4-hydroxybenzoic acid, 2gpp, d0dx8t, gsk hplc, 4-hydroxy-n-e-4-isopropylphenyl methyleneamino benzamide, 4-hydroxy-n-e-4-propan-2-ylphenyl methylideneamino benzamide, 4-hydroxy-n-1e-4-propan-2-yl phenyl methylidene benzohydrazide
PubChem CID 5331325
ChEBI CHEBI:80000
IUPAC Name 4-hydroxy-N'-[(E)-[4-(propan-2-yl)phenyl]methylidene]benzohydrazide
SMILES CC(C)C1=CC=C(\C=N\NC(=O)C2=CC=C(O)C=C2)C=C1

Specifications

Quantity 10 mg
Formula Weight 282.34
Percent Purity >99%
Chemical Name or Material GSK 4716
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.