Learn More
TARGETMOL CHEMICALS INC GSK2795039 5MG
Supplier:  TARGETMOL CHEMICALS INC T15433
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK2795039 inhibits reactive oxygen species (ROS) production and NADPH consumption. GSK2795039 decreases apoptosis. GSK2795039 is an inhibitor of NADPH oxidase 2 (NOX2) (pIC50 6 in different cell-free assays). purity: 98%
Catalog No. 50-258-1227
missing translation for 'QuantityLeft'