Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva GSTrap™ HP Columns
Shop All Cytiva Products
Click to view available options
Quantity:
1 x 5 mL
5 x 1 mL
5 x 5 mL
Description
- One-step purification of GST-tagged proteins and other glutathione S-transferases or glutathione-dependent proteins
- Glutathione coupled to Sepharose™ High Performance media
- Column material: polypropylene
- May be used with syringe, peristaltic pump or chromatography system
- Purification process preserves antigenicity and function
- Available as 1mL and 5mL columns
- Average particle size: 34μm
- Ligand: Glutathione
- Matrix: Highly cross-linked 6% agarose
- Binding capacity: >7mg/mL
- pH stability working range: 3 to 12
1mL columns
- Bed dimensions: 7 x 25mm
- Flow velocity: <4mL/min.
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
Specifications
Specifications
Bed Volume | 1 mL |
Diameter (English) Inner | 0.27 in. |
Diameter (Metric) Inner | 7 mm |
Description | GSTrap HP columns are prepacked with Glutathione Sepharose High Performance resin and provide high-resolution, one-step purifications of glutathione S-Transferase (GST) tagged proteins. |
Flow Rate | <4 mL/min. |
Material | Polypropylene |
pH Range | 3 to 12 |
Support Type | 4% Highly Cross-linked Agarose |
Content And Storage | 4°C to 30°C, 20% Ethanol |
Max. Operating Pressure | 5 bar (0.5 MPa, 70 psi) |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction