Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guaifenesin, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. p-7157524 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail

GU102, 93-14-1, C10H14O4

Spectrum™ Chemical Guaifenesin, USP is an expectorant. It helps loosen congestion in your chest and throat. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 93-14-1
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.22
MDL Number MFCD00016873
InChI Key HSRJKNPTNIJEKV-UHFFFAOYNA-N
IUPAC Name 3-(2-methoxyphenoxy)propane-1,2-diol
SMILES COC1=CC=CC=C1OCC(O)CO

Specifications

CAS Max % 100%
Loss on Drying 0.5%
Packaging Poly Bottle
Quantity 25 g
Percent Purity 98 to 102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.