Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanfacine hydrochloride, Tocris Bioscience™

Catalog No. p-4783499 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

α2A agonist

Guanfacine hydrochloride is a selective α2A-adrenoceptor agonist (Kd = 31 nM). Displays 60-fold selectivity over α2B-adrenoceptors.

Chemical Identifiers

CAS 29110-48-3
Molecular Formula C9H10Cl3N3O
Molecular Weight (g/mol) 282.549
InChI Key DGFYECXYGUIODH-UHFFFAOYSA-N
Synonym guanfacine hydrochloride, guanfacine hcl, tenex, estulic, intuniv, n-carbamimidoyl-2-2,6-dichlorophenyl acetamide hydrochloride, guanfacin hydrochloride, connexyn, n-amidino-2-2,6-dichlorophenyl acetamide hydrochloride
PubChem CID 71401
IUPAC Name N-(diaminomethylidene)-2-(2,6-dichlorophenyl)acetamide;hydrochloride
SMILES C1=CC(=C(C(=C1)Cl)CC(=O)N=C(N)N)Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 282.56
Percent Purity >99%
Chemical Name or Material Guanfacine hydrochloride
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.