Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine Hydrobromide 98.0+%, TCI America™
SDP

Catalog No. p-7130269 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 19244-98-5
Molecular Formula CH6BrN3
Molecular Weight (g/mol) 139.984
MDL Number MFCD03094062
InChI Key VQNVZLDDLJBKNS-UHFFFAOYSA-N
Synonym GABr, Guanidinium Bromide
PubChem CID 71282
IUPAC Name guanidine;hydrobromide
SMILES C(=N)(N)N.Br

Specifications

Melting Point 191°C
Color White
Quantity 1 g
Formula Weight 139.98
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Guanidine Hydrobromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.