Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine Hydrochloride, BiotechGrade, 99%, Spectrum™ Chemical
SDP

Catalog No. p-7157525 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

GU103, 50-01-1, CH5N3•HCl

Spectrum™ Chemical Guanidine Hydrochloride, BiotechGrade is considered a chaotrope and a strong denaturants utilized in physiochemical studies of protein folding. Proteins become randomly coiled at high concentrations of guanidinium chloride. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
IUPAC Name hydrogen guanidine chloride
SMILES [H+].[Cl-].NC(N)=N

Specifications

CAS Max % 100%
Melting Range 184-188°C
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 99%
Grade Biotech
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.