Learn More
Gold Biotechnology Inc Guanidine thiocyanate (Guanidine Isothiocyanate) 1 kg
SDP
Supplier:  Gold Biotechnology Inc G2101
Encompass_Preferred
During cell lysis and the extraction of DNA or RNA, denaturing enzymes such as DNase and RNase are released. To prevent DNA or RNA denaturation, the chaotropic agent, Guanadine thiocyanate is the preferred agent for use during cell lysis, and for DNA or RNA extraction. This is because of its strong ability to irreversibly inhibit the activity of DNase and RNase.
Catalog No. NC2943349
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.