Learn More
TARGETMOL CHEMICALS INC GW-493838 5MG
Supplier:  TARGETMOL CHEMICALS INC T27509
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GW-493838 is a potent adenosine A1A receptor agonist for the treatment of dyslipidemia and neuropathic pain with analgesic effects on post-herpetic neuralgia or peripheral nerve damage caused by trauma or surgery. purity: 99%
Catalog No. 50-258-0604
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.