Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GW 583340 dihydrochloride, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent dual EGFR/ErbB2 tyrosine kinase inhibitor (IC50 values are 0.01 and 0.014 μM respectively). Selectively inhibits growth of human tumor cells overexpressing EGFR and ErbB2 (IC50 values are 0.11 μM for inhibition of HN5, N87 and BT474 tumor cell lines vs. > 30 μM for inhibition of non-tumor cell line HFF). Inhibits tumor growth in vivo by ∽ 80% in a murine xenograft model following oral administration.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 671.03 |
Percent Purity | >97% |
Chemical Name or Material | GW 583340 dihydrochloride |
Safety and Handling
Recommended Storage | Desiccate at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction