Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GW 6471, Tocris Bioscience™

Catalog No. p-4817191 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

PPARα antagonist

PPARα antagonist (IC50 = 0.24 μM). Enhances the binding affinity of the PPARα ligand-binding domain to the co-repressor proteins SMRT and NCoR.

Chemical Identifiers

CAS 880635-03-0
Molecular Formula C35H36F3N3O4
Molecular Weight (g/mol) 619.685
InChI Key TYEFSRMOUXWTDN-GGGSOPIKSA-N
Synonym N-((2S)-2-(((1Z)-1-Methyl-3-oxo-3-(4-(trifluoromethyl)phenyl)prop-1-enyl)amino)-3-(4-(2-(5-methyl-2-phenyl-1, 3-oxazol-4-yl)ethoxy)phenyl)propyl)propanamide
PubChem CID 71311842
IUPAC Name N-[(2S)-3-[4-[2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)ethoxy]phenyl]-2-[[(E)-4-oxo-4-[4-(trifluoromethyl)phenyl]but-2-en-2-yl]amino]propyl]propanamide
SMILES CCC(=O)NCC(CC1=CC=C(C=C1)OCCC2=C(OC(=N2)C3=CC=CC=C3)C)NC(=CC(=O)C4=CC=C(C=C4)C(F)(F)F)C

Specifications

Quantity 10 mg
Formula Weight 619.67
Percent Purity >98%
Chemical Name or Material GW 6471
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.