Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GW 843682X, Tocris Bioscience™

Catalog No. p-4784011 Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective inhibitor of PLK1 and PLK3

Selective inhibitor of polo-like kinase 1 (PLK1) and polo-like kinase 3 (PLK3) (IC50 values are 2.2 and 9.1 nM respectively). Displays > 100-fold selectivity over ∽30 other kinases tested including cdk1 and cdk2. Inhibits proliferation of most tumor cells in vitro and is selective over normal diploid fibroblasts.

Chemical Identifiers

CAS 660868-91-7
Molecular Formula C22H18F3N3O4S
Molecular Weight (g/mol) 477.458
InChI Key JSKUWFIZUALZLX-UHFFFAOYSA-N
Synonym 5-5,6-dimethoxy-1h-benzimidazol-1-yl-3-2-trifluoromethyl-benzyl oxy thiophene-2-carboxamide, 5-5,6-dimethoxy-1h-benzimidazol-1-yl-3-2-trifluoromethyl benzyl oxy thiophene-2-carboxamide, 5-5,6-dimethoxybenzimidazol-1-yl-3-2-trifluoromethyl phenyl methoxy thiophene-2-carboxamide, kinome_3462, thiophene deriv., 20, bmcl191018 compound 2, gw843682x hplc , solid, 3-2-trifluoromethyl benzyloxy-5-5,6-dimethoxy-1h-benzo d imidazol-1-yl thiophene-2-carboxamide, 5-5,6-dimethoxy-1,3-benzodiazol-1-yl-3-2-trifluoromethyl phenyl methoxy thiophene-2-carboxamide, 5-5,6-dimethoxy-1-benzimidazolyl-3-2-trifluoromethyl phenyl methoxy-2-thiophenecarboxamide
PubChem CID 9826308
IUPAC Name 5-(5,6-dimethoxybenzimidazol-1-yl)-3-[[2-(trifluoromethyl)phenyl]methoxy]thiophene-2-carboxamide
SMILES COC1=C(C=C2C(=C1)N=CN2C3=CC(=C(S3)C(=O)N)OCC4=CC=CC=C4C(F)(F)F)OC

Specifications

Quantity 1 mg
Formula Weight 477.46
Percent Purity >98%
Chemical Name or Material GW 843682X
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.