Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H 89 dihydrochloride, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Description
H 89 dihydrochloride is a protein kinase A inhibitor that also inhibits several other kinases (IC50 values are 80, 120, 135, 270, 2600 and 2800 nM for S6K1, MSK1, PKA, ROCKII, PKBα and MAPKAP-K1b). Exhibits antinociceptive activity. Enhances survival and clonogenicity of dissociated human ESCs through ROCK inhibition.
Specifications
Specifications
Quantity | 1 mg |
Formula Weight | 519.28 |
Percent Purity | >99% |
Chemical Name or Material | H 89 dihydrochloride |
Safety and Handling
Recommended Storage | Store at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction