Learn More
Selleck Chemical LLC H2DCFDA-25mg
Supplier:  Selleck Chemical LLC S968725MG
Encompass_Preferred
H2DCFDA (DCFH-DA, 2',7'-Dichlorodihydrofluorescein diacetate) is a cell-permeable probe used to detect intracellular reactive oxygen species (ROS) production. It is de-esterified intracellularly and turns to highly fluorescent 2′,7′-dichlorofluorescein upon oxidation. It is used in sensitive and rapid quantitation of oxygen-reactive species in response to oxidative metabolism; microplate assay for detecting oxidative products in phagocytic cells, and quantitative multiwell myeloid differentiation assay.
Catalog No. 50-238-9620
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.