Learn More
NEOGEN Half Fraser (Demi-Fraser) Broth (ISO), 500 g
Supplier: NEOGEN 700002945
Half Fraser (Demi-Fraser) Broth is used with ferric ammonium citrate for the selective enrichment of Listeria species and is not intended for use in the diagnosis of disease or other conditions in humans. Primary enrichment broth for the isolation of Listeria spp. formulated according to ISO 11290. The selective components acriflavine and nalidixic acid are blended into the base powder and the ferric ammonium citrate is added to the tempered broth after sterilization. Half Fraser (Demi-Fraser) Broth, NCM0001 was developed to give improved results for the isolation of Listeria spp. with both traditional and ELISA methods. As with the sister product, Fraser Broth, NCM0050 the selective components acriflavine and nalidixic acid are blended into the base powder and the Ferric Ammonium Citrate is added to the tempered broth after autoclaving.For more information visit https://www.neogen.com/solutions/Microbiology/half-fraser-broth
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.