Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Halosulfuron-methyl 98.0+%, TCI America™
SDP

Catalog No. p-7131012 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 100784-20-1
Molecular Formula C13H15ClN6O7S
Molecular Weight (g/mol) 434.808
MDL Number MFCD01631160
InChI Key FMGZEUWROYGLAY-UHFFFAOYSA-N
Synonym Methyl 3-Chloro-5-[[3-(4,6-dimethoxypyrimidin-2-yl)ureido]sulfonyl]-1-methyl-1H-pyrazole-4-carboxylate, 3-Chloro-5-[[3-(4,6-dimethoxypyrimidin-2-yl)ureido]sulfonyl]-1-methyl-1H-pyrazole-4-carboxylic Acid Methyl Ester
PubChem CID 91763
ChEBI CHEBI:81750
IUPAC Name methyl 3-chloro-5-[(4,6-dimethoxypyrimidin-2-yl)carbamoylsulfamoyl]-1-methylpyrazole-4-carboxylate
SMILES CN1C(=C(C(=N1)Cl)C(=O)OC)S(=O)(=O)NC(=O)NC2=NC(=CC(=N2)OC)OC

Specifications

Melting Point 174°C
Quantity 1 g
UN Number 3077
Formula Weight 434.81
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Halosulfuron-methyl
RTECSNumber UQ6392606
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.