Learn More
Uline Hard Hat Face Shield Adapter | | 1 EA
Supplier:  Uline S24404
Encompass_Preferred
Spring-mounted aluminum adapter pivots up and down. Easily attach to hard hat then clip on shield. Use with Hard Hat Face Shield sold separately. Use with Standard Vented and Bump Cap Hard Hats sold separately.
Catalog No. NC2386289
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.