Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HARLECO™ Alcohol, 70%, MilliporeSigma™
GSA_VA

Catalog No. 23794203 Shop All MilliporeSigma Products
Click to view available options
Quantity:
Packaging:

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS

64-17-5

Molecular Formula

CH4O

Molecular Weight (g/mol)

32.04

MDL Number

MFCD00004595

InChI Key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Synonym

methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit

PubChem CID

702

ChEBI

CHEBI:16236

IUPAC Name

methanol

SMILES

CO

Specifications

Physical Form

Liquid

Packaging

Poly bottle

Quantity

4 L

Assay Percent Range

70%

Linear Formula

C2H5OH

Formula Weight

46.8

Chemical Name or Material

Alcohol, HARLECO™

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.