Learn More
Fisherbrand™ Hazardous Chemical Dispensers
SCI_ED
Bottle-top dispensers for use with hazardous and corrosive chemicals.
 View more versions of this product
Catalog No. S29125
This item is not returnable. View return policy
Description
Description
  • Constructed entirely of PTFE with totally inert pouring surfaces—even the neck-sealing ring is encapsulated in PTFE
  • Incorporated breather vent allows for direct and controlled pourings
  • Captured PTFE closure ensures contaminant-free contents when not in use
  • Fit standard reagent bottles with neck sizes of GL32, 38, and 45
Specifications
Specifications
Neck size: 38 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.