Learn More
Gold Biotechnology Inc HB101 Chemically Competent E. coli Cells 5x50 μl
SDP
Supplier:  Gold Biotechnology Inc CC1505X50
Encompass_Preferred
GoldBio’s HB101 chemically competent E. coli cells allow you to obtain high transformation efficiency in applications such cloning and sub-cloning. Our E. coli HB101 proceeds from the K12 x B hybrid strain. HB101 strain also contains the recA13 mutation useful in the insert stability and minimization of recombination. E. coli HB101 also has the hsdS20(rB-mB-) genotype that prevents the cleavage of cloned DNA by endogenous restriction enzymes.
Catalog No. NC2942854
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.