Learn More
Gibco™ HBSS (10X), calcium, magnesium, phenol red

Description
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.
This HBSS is modified as follows:
- With: Calcium, Magnesium, Glucose, Phenol Red
- Without: Sodium Bicarbonate
The complete formulation is available.
Concentrated forms of Gibco™ HBSS require pH adjustment and filtration at the time of preparation (see protocol for details).
Specifications
Specifications
Additive | Calcium and Magnesium |
Classification | Animal Origin-Free |
Content And Storage | Storage conditions: 15°C to 30°C Shipping conditions: Ambient Shelf life: 36 months from date of manufacture |
Dilution | 10X |
Product Line | Gibco™ |
Product Type | Hanks' Balanced Salt Solution (HBSS) |
Quantity | 500 mL |
Solution Type | Hank's Balanced Salt |
Green Features | Sustainable packaging |
Osmolality | 2500 to 3000 mOsm/kg |
Show More |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.