Learn More
Medchemexpress LLC HY-15064 100mg , HC-030031 CAS:349085-38-7 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY15064100MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-15064 100mg HC-030031 CAS:349085-38-7 Formula:C18H21N5O3 TRPA1 Purity:98% HC-030031 is a potent and selective of TRPA1 inhibitor, which antagonizes AITC- and formalin-evoked calcium influx with IC 50 s of 6.2±0.2 and 5.3±0.2 μM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3549
missing translation for 'QuantityLeft'