Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HC 030031, Tocris Bioscience™
Shop All R&D Systems Products

Click to view available options
Quantity:
10 mg
50 mg
Description
Selective TRPA1 channel blocker that antagonizes AITC- and formalin-evoked calcium influx (IC50 values are 6.2 and 5.3 μM respectively). Does not block currents mediated by TRPV1, TRPV3, TRPV4, hERG or NaV1.2 channels. Inhibits AITC- and formalin-induced flinching in vivo.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 355.39 |
Percent Purity | >99% |
Chemical Name or Material | HC 030031 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction