Learn More
TARGETMOL CHEMICALS INC HCAI/II-IN-6 5MG
Supplier:  TARGETMOL CHEMICALS INC T61745
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. hCAI/II-IN-6 is a selective and orally active inhibitor of human carbonic anhydrase (CA). hCAI/II-IN-6 inhibited hCA I hCA II hCA VII and hCA XII with Ki values of 220 4.9 6.5 and >50000 nM. hCAI/II-IN-6 showed anticonvulsant activity in vivo. hCAI/II-IN-6 can be used to study epilepsy. hCAI/II-IN-6 showed anticonvulsant activity in vivo. hCAI/II-IN-6 can be used to study epilepsy. purity: 97%
Catalog No. 50-257-9504