Learn More
TARGETMOL CHEMICALS INC HCAII-IN-9 5MG
Supplier:  TARGETMOL CHEMICALS INC T73177
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. hCAII-IN-9 is a potent carbonic anhydrase inhibitor that inhibits hCA II hCA IX and hCA XII with IC50s of 1.18 uM and 0.17 uM and 2.99 uM respectively. hCAII-IN-9 is not blood-brain barrier permeable. purity: 98%
Catalog No. 50-257-9219
missing translation for 'QuantityLeft'