Learn More
Research Products International Corp Heat Sealable Poly Bag, 4 mm Thick, 23 x 23 cm, 100 per Case
SDP
Supplier:  Research Products International Corp 286662C
Encompass
Heat Sealable Polyethylene Bags are transparent and specially designed of use in all hybridization applications. Ideal for quickly sealing solutions with a bag sealer. Bags are 4 mm thick. Poly Bags can be used with an Electronic Bag Sealer also available from RPI.
Catalog No. 50-212-876
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.