Learn More
Eurofins Discoverx PathHunter HEK 293 SCTR β-Arrestin Cell Line, 2 vials. (93-0237C1)
Supplier:  Eurofins Discoverx 930237C1
Encompass
The PathHunter SCTR cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. secretin) based activation of SCTR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged SCTR and EA-tagged β-Arrestin-2. Activation of SCTR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0038
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.