Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Helix pomatia Copper-Metallothionein Rabbit anti-Mollusca, Polyclonal, Invitrogen™
Description
This antibody has been tested in direct-ELISA
Helix pomatia Copper-Metallothionein Polyclonal antibody specifically detects Helix pomatia Copper-Metallothionein in Mollusca samples. It is validated for Western BlotSpecifications
Helix pomatia Copper-Metallothionein | |
Polyclonal | |
Unconjugated | |
Cu-MT | |
Recombinant Roman snail Copper-metallothionein protein (1-64aa) | |
100 μL | |
Primary | |
-20°C or -80°C if preferred | |
Liquid |
Western Blot | |
4.355 mg/mL | |
10mM PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Rabbit | |
Protein G | |
RUO | |
Mollusca | |
Antibody | |
IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?