Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heparin sodium salt, Tocris Bioscience™

Catalog No. 2812100 Shop All R&D Systems Products
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Anticoagulant

Minimum activity: > 150 I.U./mg. Glycosaminoglycan that behaves as an anticoagulant. Binds with high affinity to antithrombin III (AT-III).

Chemical Identifiers

CAS 9041-08-1
MDL Number MFCD00081689

Specifications

Quantity 100 mg
Percent Purity >98%
Chemical Name or Material Heparin sodium salt
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.