Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heparin sodium, Thermo Scientific Chemicals

Catalog No. p-140388
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle

Heparin Sodium Salt, CAS # 9041-08-1, is derived from porcine intestinal mucosa and shows blood anti-coagulant activity.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Heparin sodium salt displays anti-coagulant activity. It can also efficiently inhibit mesenchymal cell proliferation and promotes the proliferation of primary epithelial cells
  • This compound can bind to antithrombin III and inhibit coagulation proteases

Applications

  • In in vitro studies, it is used as the main component of serum-free media to prevent cell aggregation
  • Heparin sodium salt can also induce the growth of human embryonic stem cells in a defined serum-free medium, acting as a cofactor for a basic fibroblast growth factor (FGF-2)
  • It is also applied in laboratory studies related to stem cell differentiation and growth
  • In in vitro studies this compound exhibits cytotoxic action inducing cancer cell death by interfering with transcription factor activity

Chemical Identifiers

CAS 9041-08-1
MDL Number MFCD00081689

Specifications

Absorbance ≤0.2% as nucleotides, 0.2 max. (0.4% in water) at 260nm, Protein and nucleotide impurities:, Protein and nucleotide impurities: at 0nm
Color White
pH Inclusive between 5.5 (1% soln.)
Ignition Residue Inclusive between 30%
Loss on Drying ≤8% (at 60°C for 4 hr.)
Infrared Spectrum Pass Test
Packaging Glass Bottle
Quantity 1 g
Merck Index 15, 4688
Percent Purity 150 I.U./mg min.
Concentration or Composition (by Analyte or Components) ≥150 I.U./mg
Physical Form Powder
Chemical Name or Material Heparin Sodium Salt
Show More Show Less
Hazard Category HYGROSCOPIC
RTECSNumber MI0850000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.