Learn More
Hepsin Monoclonal Antibody (2D5), Functional Grade, In Vivo GOLD™, Leinco Technologies™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc H4400100MG

Description
Specificity: Clone 2D5 recognizes Human hepsin. It binds to a separate epitope than clone 3H10.1. These clones do not interfere with one another. Endotoxin Level: <1.0 EU/mg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.
Hepsin Monoclonal antibody specifically detects Hepsin in Human samples. It is validated for ELISA, Flow Cytometry, Functional Assay, Western BlotSpecifications
Hepsin | |
Monoclonal | |
≥5.0 mg/mL | |
PBS with no preservative; pH 7.2-7.4 | |
P05981 | |
Mouse | |
Protein A/G | |
RUO | |
3249 | |
4°C short term, -80°C long term | |
Liquid |
ELISA, Flow Cytometry, Functional Assay, Western Blot | |
2D5 | |
Functional Grade | |
HPN | |
hepsin; hepsin (transmembrane protease, serine 1); Hlb320; HPN; Serine protease hepsin; Serine protease hepsin catalytic chain; Serine protease hepsin non-catalytic chain; testicular tissue protein Li 85; TMPRSS1; Transmembrane protease serine 1 | |
Human hepsin protein | |
100 mg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.