Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptacosanoic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7130834 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g
5 g

Chemical Identifiers

CAS 7138-40-1
Molecular Formula C27H54O2
Molecular Weight (g/mol) 410.73
MDL Number MFCD00014029
InChI Key VXZBFBRLRNDJCS-UHFFFAOYSA-N
PubChem CID 23524
ChEBI CHEBI:78710
IUPAC Name heptacosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O

Specifications

Melting Point 86°C
Color White-Yellow
Quantity 100 mg
Formula Weight 410.73
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Heptacosanoic Acid
EINECSNumber (2)-0608
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.