Learn More
Ampco Safety Tools Hex Key Kit, 10 Piece Metric

Ampco Safety Tools Hex Key Kit is non-sparking, non-magnetic and corrosion resistant
Supplier: Ampco Safety Tools M42M
Description
- Sizes 1.5mm, 2mm, 3mm, 4mm, 5mm, 6mm, 7mm, 8mm, 9mm and 10mm
- Made in USA
Certifications
FM Approved
Warranty and Services
Lifetime Warranty with proper use. If a tool breaks or fails to perform under normal use aor service, it will be repaired or eplaced at no charge - exceptions include defects resulting from misuse, abuse or normal service wear.
Specifications
Electronics, batteries, MRI, and precision magnetic equipment | |
Beryllium Copper | |
Non-Sparking, Non-Magnetic, Corrosion Resistant Applications; Flammable or combustible vapors, liquids, dusts or residues; Corrosive environments like laboratories, acid, salt water and chemical processing | |
430 g |
228.6 mm | |
Bronze | |
1 Ea. | |
228.6 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.