Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecamethylcyclooctasiloxane 96.0+%, TCI America™
SDP

Catalog No. p-7130961 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 mL
5 mL

Chemical Identifiers

CAS 556-68-3
Molecular Formula C16H48O8Si8
Molecular Weight (g/mol) 593.232
InChI Key XKJMJYZFAWYREL-UHFFFAOYSA-N
PubChem CID 11170
IUPAC Name 2,2,4,4,6,6,8,8,10,10,12,12,14,14,16,16-hexadecamethyl-1,3,5,7,9,11,13,15-octaoxa-2,4,6,8,10,12,14,16-octasilacyclohexadecane
SMILES C[Si]1(O[Si](O[Si](O[Si](O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)(C)C)(C)C)(C)C)C

Specifications

Color Yellow
Boiling Point 168°C
Quantity 1 mL
Formula Weight 593.23
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexadecamethylcyclooctasiloxane
EINECSNumber (7)-0475
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.