Learn More
Electron Microscopy Sciences Veco Hexagonal 200 mesh Copper Grid
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter, 0.8 mil thickness.
Grid: Copper