Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hexakis(1H,1H,omegaH-perfluoroalkoxy)phosphazene, mass spectrometry standard, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Specifications
Specifications
Density | 1.8000g/mL |
Packaging | Glass bottle |
Refractive Index | 1.3490 to 1.3570 |
Quantity | 250 mg |
Specific Gravity | 1.8 |
Synonym | ultramark1621, ultramark 1621, hexa 2 h? methyl-1,3,5,2??,4??,6??-triazatriphosphinine |
Formula Weight | 2120 |
Chemical Name or Material | Hexakis(1H, 1H, ωH-perfluoroalkoxy)phosphazene |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction